siRNA Details
Gene Symbol: 9430096G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020526.1 | - | 1 | 226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721953.1 | - | 1 | 223 | Chr. 2 - 170318632 - 170319230 [-] on Build GRCm38 |
