siRNA Details
Gene Symbol: 9430093N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020520.1 | - | 2 | 418 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721935.1 | - | 2 | 414 | Chr. 2 - 169234993 - 169237780 [+] on Build GRCm38 |
