siRNA Details
Gene Symbol: E130114A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021388.1 | - | 1 | 808 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB541403.1 | - | 1 | 118 | Chr. X - 85733876 - 85734110 [-] on Build GRCm38 |
