siRNA Details
Gene Symbol: 9330179D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040273.1 | 3 | 929 | |||
NR_040274.1 | 3 | 911 | ||||
NR_040275.1 | 2 | 723 | ||||
GenBank mRNA | AK020388.1 | - | 2 | 725 | ||
AK043994.1 | - | 3 | 911 |
