siRNA Details
Gene Symbol: 9330179D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040273.1 | 3 | 731 | |||
NR_040274.1 | 3 | 713 | ||||
NR_040275.1 | 2 | 525 | ||||
GenBank mRNA | AK020388.1 | - | 2 | 527 | ||
AK043994.1 | - | 3 | 713 | |||
CX238860.1 | - | 3 | 604 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721457.1 | - | 2 | 523 | Chr. 6 - 127149628 - 127211952 [+] on Build GRCm38 |
