siRNA Details
Gene Symbol: 0710001D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002941.1 | - | 1 | 965 | ||
AK006324.1 | - | 1 | 721 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 0710001D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002941.1 | - | 1 | 965 | ||
AK006324.1 | - | 1 | 721 |