siRNA Details
Gene Symbol: 1700015G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005994.1 | - | 4 | 271 | ||
AV042221.2 | - | 2 | 154 | |||
AV042559.2 | - | 2 | 160 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700015G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005994.1 | - | 4 | 271 | ||
AV042221.2 | - | 2 | 154 | |||
AV042559.2 | - | 2 | 160 |