siRNA Details
Gene Symbol: 1700015I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005997.1 | - | 1 | 312 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV041027.2 | - | 1 | 238 | Chr. 8 - 35708587 - 35708863 [-] on Build GRCm38 |
