siRNA Details
Gene Symbol: Gm15774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143626.1 | - | 308 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY732403.1 | - | 2 | 307 | Chr. 7 - 120874029 - 120875298 [-] on Build GRCm38 |
