siRNA Details
Gene Symbol: BC051408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC051408.1 | - | 25 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ949344.1 | - | 20 | Chr. 18 - 28711861 - 28725979 [+] on Build GRCm38 |
