siRNA Details
Gene Symbol: 4930528J11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015928.1 | - | 1 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715487.1 | - | 1 | 487 | Chr. 6 - 29915311 - 29915926 [+] on Build GRCm38 |
