siRNA Details
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139055.1 | - | 1100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF551415.1 | - | 1 | 71 | Chr. 3 - 103007165 - 103008013 [+] on Build GRCm38 |
