siRNA Details
Gene Symbol: Gm32635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135920.1 | - | 3 | 674 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719847.1 | - | 3 | 668 | Chr. 12 - 106095283 - 106098971 [-] on Build GRCm38 |
