siRNA Details
Gene Symbol: Lrrc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144081.1 | - | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ068761.1 | - | 1 | 74 | Chr. 12 - 72520091 - 72530575 [+] on Build GRCm38 |
