siRNA Details
Gene Symbol: 4933412E12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038025.1 | 4 | 1055 | |||
NR_038026.1 | 3 | 842 | ||||
GenBank mRNA | AK016788.1 | - | 4 | 1030 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933412E12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038025.1 | 4 | 1055 | |||
NR_038026.1 | 3 | 842 | ||||
GenBank mRNA | AK016788.1 | - | 4 | 1030 |