siRNA Details
Gene Symbol: 1700104L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108033.1 | 2, 3 | 254 | |||
GenBank mRNA | AK007120.1 | - | 2, 3 | 255 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707200.1 | - | 2, 3 | 255 | Chr. 12 - 54233615 - 54242315 [+] on Build GRCm38 |
