siRNA Details
Gene Symbol: 1700008B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005752.1 | - | 1 | 325 | ||
AK007127.1 | - | 1 | 352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707205.1 | - | 1 | 352 | Chr. 5 - 121835195 - 121835707 [+] on Build GRCm38 |
