siRNA Details
Gene Symbol: Gm33374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039957.1 | - | 3 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634637.1 | - | 3 | 210 | Chr. 10 - 19716240 - 19718703 [-] on Build GRCm38 |
