siRNA Details
Gene Symbol: 8430439C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018478.1 | - | 1 | 313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720612.1 | - | 1 | 307 | Chr. 9 - 6911593 - 6912371 [-] on Build GRCm38 |
