siRNA Details
Gene Symbol: 8430437O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018474.1 | - | 1 | 384 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720608.1 | - | 1 | 384 | Chr. 1 - 172184427 - 172185074 [+] on Build GRCm38 |
