siRNA Details
Gene Symbol: Lppos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037954.1 | 2 | 596 | |||
GenBank mRNA | AK004252.1 | - | 2 | 384 | ||
AK039760.1 | - | 402 | ||||
BC037367.1 | - | 1 | 290 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lppos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037954.1 | 2 | 596 | |||
GenBank mRNA | AK004252.1 | - | 2 | 384 | ||
AK039760.1 | - | 402 | ||||
BC037367.1 | - | 1 | 290 |