siRNA Details
Gene Symbol: 2310009A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003016.1 | - | 2 | 186 | ||
AK009244.1 | - | 2 | 193 | |||
BC037369.1 | - | 2 | 160 | |||
BC100521.1 | - | 2 | 185 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310009A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003016.1 | - | 2 | 186 | ||
AK009244.1 | - | 2 | 193 | |||
BC037369.1 | - | 2 | 160 | |||
BC100521.1 | - | 2 | 185 |