siRNA Details
Gene Symbol: Gm19619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040428.1 | 3 | 421 | |||
GenBank mRNA | AK050091.1 | - | 3 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB653650.1 | - | 3 | 422 | Chr. 5 - 91280493 - 91283076 [-] on Build GRCm38 |
