siRNA Details
Gene Symbol: Gm33602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078946.1 | - | 3 | 1339 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV377002.2 | - | 2 | 398 | Chr. X - 9179464 - 9187404 [-] on Build GRCm38 |
