siRNA Details
Gene Symbol: Gm37896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044741.1 | - | 3 | 590 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642797.1 | - | 3 | 590 | Chr. 1 - 187447877 - 187459603 [+] on Build GRCm38 |
