siRNA Details
Gene Symbol: 9130017K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020265.1 | - | 1 | 62 | ||
AK043616.1 | - | 2 | 2899 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130017K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020265.1 | - | 1 | 62 | ||
AK043616.1 | - | 2 | 2899 |