siRNA Details
Gene Symbol: B930036N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154483.1 | - | 490 | |||
AK171461.1 | - | 1 | 502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD560072.1 | - | 1 | 298 | Chr. 1 - 171502427 - 171502960 [-] on Build GRCm38 |
