siRNA Details
Gene Symbol: Gm11732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045779.1 | - | 4 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB643605.1 | - | 4 | 653 | Chr. 11 - 117426178 - 117437591 [+] on Build GRCm38 |
