siRNA Details
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014203.1 | - | 1 | 315 | ||
AK033622.1 | - | 1 | 1826 | |||
AK157606.1 | - | 1 | 1859 | |||
AK169176.1 | - | 1 | 2635 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014203.1 | - | 1 | 315 | ||
AK033622.1 | - | 1 | 1826 | |||
AK157606.1 | - | 1 | 1859 | |||
AK169176.1 | - | 1 | 2635 |