siRNA Details
Gene Symbol: 4931428A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015556.1 | - | 1 | 664 | ||
AK016478.1 | - | 1 | 633 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715106.1 | - | 1 | 662 | Chr. 1 - 90758619 - 90759355 [+] on Build GRCm38 |
