siRNA Details
Gene Symbol: 4930512B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033573.1 | 5 | 627 | |||
GenBank mRNA | AK015558.1 | - | 5 | 637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715108.1 | - | 5 | 637 | Chr. 12 - 69790335 - 69840477 [+] on Build GRCm38 |
