siRNA Details
Gene Symbol: 4930512B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033573.1 | 4 | 371 | |||
GenBank mRNA | AK015558.1 | - | 4 | 381 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715108.1 | - | 4 | 381 | Chr. 12 - 69790335 - 69840477 [+] on Build GRCm38 |
