siRNA Details
Gene Symbol: 4930512B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033573.1 | 7 | 941 | |||
GenBank mRNA | AK015558.1 | - | 7 | 951 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930512B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033573.1 | 7 | 941 | |||
GenBank mRNA | AK015558.1 | - | 7 | 951 |