siRNA Details
Gene Symbol: Son | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003921.1 | - | 180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY704175.1 | - | 1 | 180 | Chr. 16 - 91679067 - 91679460 [+] on Build GRCm38 |
