siRNA Details
Gene Symbol: Tnfrsf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048111.1 | - | 1 | 540 | ||
AK081533.1 | - | 1 | 540 | |||
BC076592.1 | - | 6 | 2921 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK042818.1 | - | 1632 | Chr. - - [-] on Build |
