siRNA Details
Gene Symbol: Gm10832 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136598.1 | - | 2 | 323 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720934.1 | - | 2 | 323 | Chr. 16 - 11031956 - 11036420 [+] on Build GRCm38 |
