siRNA Details
Gene Symbol: Gm34639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136628.1 | - | 3 | 299 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720988.1 | - | 3 | 299 | Chr. 13 - 28890390 - 28897945 [-] on Build GRCm38 |
