siRNA Details
Gene Symbol: 9130015G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035625.1 | - | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB628205.1 | - | 2 | 233 | Chr. 7 - 49123646 - 49159819 [-] on Build GRCm38 |
