siRNA Details
Gene Symbol: 5330413P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089122.2 | - | 2 | 538 | ||
AK134994.1 | - | 2 | 441 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718980.1 | - | 2 | 436 | Chr. 2 - 131845188 - 131847656 [-] on Build GRCm38 |
