siRNA Details
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038001.1 | 2 | 114 | |||
GenBank mRNA | AK006406.1 | - | 2 | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706463.1 | - | 2 | 112 | Chr. 1 - 73016038 - 73025498 [+] on Build GRCm38 |
