siRNA Details
Gene Symbol: Gm29680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144747.1 | - | 6 | 577 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734286.1 | - | 6 | 575 | Chr. 13 - 90181376 - 90217141 [-] on Build GRCm38 |
