siRNA Details
Gene Symbol: 2700001H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019195.1 | - | 1 | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE648109.1 | - | 1 | 424 | Chr. 10 - 126971616 - 126972074 [-] on Build GRCm38 |
