siRNA Details
Gene Symbol: 5930403N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031090.1 | - | 1 | 949 | ||
AK040320.1 | - | 1 | 593 | |||
BC112909.1 | - | 1 | 1104 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177177.1 | - | 949 | Chr. - - [-] on Build |
