siRNA Details
Gene Symbol: 6530437J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020101.1 | - | 1, 2 | 363 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719281.1 | - | 1, 2 | 363 | Chr. 7 - 44970427 - 44974175 [+] on Build GRCm38 |
