siRNA Details
Gene Symbol: 3110006O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014014.1 | - | 1 | 47 | ||
BC067033.1 | - | 1 | 32 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF530849.1 | - | 1 | 32 | Chr. 13 - 78171630 - 78179624 [+] on Build GRCm38 |
