siRNA Details
Gene Symbol: 4930511M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015762.1 | - | 2 | 111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715304.1 | - | 2 | 111 | Chr. 3 - 92102573 - 92142736 [-] on Build GRCm38 |
