siRNA Details
Gene Symbol: 4930529N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015940.1 | - | 2 | 343 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715503.1 | - | 2 | 340 | Chr. 13 - 35167146 - 35167802 [-] on Build GRCm38 |
