siRNA Details
Gene Symbol: 4930599N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045813.1 | 4 | 796 | |||
GenBank mRNA | AK016417.1 | - | 4 | 796 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930599N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045813.1 | 4 | 796 | |||
GenBank mRNA | AK016417.1 | - | 4 | 796 |