siRNA Details
Gene Symbol: 4930599N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045813.1 | 4 | 670 | |||
GenBank mRNA | AK016417.1 | - | 4 | 670 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716068.1 | - | 4 | 669 | Chr. 6 - 39118473 - 39148280 [+] on Build GRCm38 |
