siRNA Details
Gene Symbol: 2810017D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012752.1 | - | 1 | 199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB194530.1 | - | 1 | 129 | Chr. 4 - 130709741 - 130710509 [+] on Build GRCm38 |
